Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cat-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267080.3 | NP_001254009.1 | 6 - 7 | 1333 | 73 | |
NM_001306554.2 | NP_001293483.1 | 6 - 7 | 1264 | 73 | ||
NM_001383774.1 | 5 - 6 | 1970 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |