Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ifb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383849.1 | 2 - 3 | 415 | 77 | ||
NM_062736.4 | NP_495137.1 | 2 - 3 | 308 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |