Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ire-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267206.3 | NP_001254135.1 | 5 - 6 | 826 | 115 | |
GenBank mRNA | AF435952.1 | - | 4 - 5 | 689 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |