Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ooc-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063515.3 | NP_495916.1 | 2 - 3 | 212 | 149 | |
NM_063516.5 | NP_495917.1 | 2 - 3 | 185 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |