Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gbh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063904.7 | NP_496305.1 | 6 - 7 | 1098 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |