Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cap-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063935.9 | NP_496336.1 | 5 - 6 | 489 | 80 | |
GenBank mRNA | Z18854.1 | - | 333 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |