Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cap-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063935.9 | NP_496336.1 | 6 - 7 | 644 | 102 | |
GenBank mRNA | Z18854.1 | - | 488 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |