Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: imb-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322761.3 | NP_001309574.1 | 1 - 2 | 76 | 65 | |
NM_064586.4 | NP_496987.1 | 1 - 2 | 76 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |