Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: unc-52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027317.8 | NP_001022488.1 | 3 - 4 | 771 | 65 | |
NM_001267510.3 | NP_001254439.1 | 3 - 4 | 770 | 65 | ||
NM_001267511.3 | NP_001254440.1 | 3 - 4 | 680 | 65 | ||
NM_001267513.3 | NP_001254442.1 | 3 - 4 | 771 | 65 | ||
NM_001267514.4 | NP_001254443.1 | 3 - 4 | 680 | 65 | ||
NM_001267515.3 | NP_001254444.1 | 3 - 4 | 680 | 65 | ||
NM_001267516.3 | NP_001254445.1 | 3 - 4 | 680 | 65 | ||
NM_001267517.3 | NP_001254446.1 | 3 - 4 | 680 | 65 | ||
NM_001350388.3 | NP_001337294.1 | 13 | 3047 | 65 | ||
NM_001377905.1 | NP_001364664.1 | 13 - 14 | 3207 | 65 | ||
NM_064643.10 | NP_497044.3 | 3 - 4 | 680 | 65 | ||
NM_064644.6 | NP_497045.1 | 3 - 4 | 770 | 65 | ||
NM_064645.6 | NP_497046.1 | 3 - 4 | 771 | 65 | ||
NM_064646.7 | NP_497047.1 | 3 - 4 | 771 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |