Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: polh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065079.3 | NP_497480.2 | 2 - 3 | 292 | 82 | |
GenBank mRNA | AB244413.1 | - | 2 - 3 | 275 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |