Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxa-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065112.2 | NP_497513.1 | 1 - 2 | 139 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |