Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdka-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083180.3 | NP_001076649.1 | 6 - 7 | 881 | 64 | |
NM_001083181.5 | NP_001076650.1 | 7 - 8 | 1106 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |