Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: set-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027625.4 | NP_001022796.1 | 3 - 4 | 612 | 70 | |
NM_001379764.2 | NP_001367441.1 | 3 - 4 | 626 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |