Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066346.6 | NP_498747.2 | 1 - 2 | 106 | 138 | |
GenBank mRNA | AF104917.1 | - | 1 - 2 | 109 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |