Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fli-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066512.6 | NP_498913.2 | 12 - 13 | 3802 | 104 | |
GenBank mRNA | U01183.1 | - | 13 - 14 | 3835 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |