Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001372642.1 | NP_001359524.1 | 6 - 7 | 2489 | 148 | |
NM_066666.5 | NP_499067.3 | 6 - 7 | 2181 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |