Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mog-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066811.9 | NP_499212.1 | 4 - 5 | 2369 | 95 | |
GenBank mRNA | AF120269.1 | - | 4 - 5 | 2384 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |