Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nud-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067348.7 | NP_499749.1 | 1 - 2 | 108 | 75 | |
GenBank mRNA | AF164431.1 | - | 1 - 2 | 105 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |