Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047473.4 | NP_001040938.1 | 4 - 5 | 864 | 77 | |
NM_001047474.3 | NP_001040939.1 | 2 - 3 | 300 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |