Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bre-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083199.4 | NP_001076668.1 | 3 - 4 | 634 | 134 | |
NM_001083200.3 | NP_001076669.1 | 4 - 5 | 984 | 134 | ||
NM_001083201.4 | NP_001076670.1 | 5 - 6 | 858 | 134 | ||
NM_069162.9 | NP_501563.1 | 4 - 5 | 689 | 134 | ||
NM_069163.5 | NP_501564.1 | 4 - 5 | 792 | 134 | ||
GenBank mRNA | AM231683.1 | - | 4 - 5 | 727 | 134 | |
AM231684.1 | - | 4 - 5 | 682 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |