Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: del-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268482.3 | NP_001255411.1 | 7 - 8 | 1085 | 151 | |
NM_001380416.1 | NP_001367239.1 | 3 - 4 | 667 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |