Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: thk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268549.3 | NP_001255478.1 | 1 - 2 | 119 | 129 | |
NM_001383184.1 | 8 - 9 | 1165 | 129 | |||
GenBank mRNA | AY523517.1 | - | 2 - 3 | 161 | 129 |
Gene Symbol: rad-51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028294.4 | NP_001023465.1 | 9 - 10 | 1309 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |