Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gly-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047571.4 | NP_001041036.2 | 3 - 4 | 590 | 99 | |
NM_001047572.4 | NP_001041037.1 | 3 - 4 | 347 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |