Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269027.3 | NP_001255956.2 | 4 - 5 | 414 | 63 | |
NM_001269028.2 | NP_001255957.2 | 3 - 4 | 444 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |