Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nas-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047437.2 | NP_001040902.1 | 6 - 7 | 891 | 151 | |
NM_001047438.2 | NP_001040903.1 | 2 - 3 | 117 | 151 | ||
NR_132470.1 | 6 - 7 | 613 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |