Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtm-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071637.6 | NP_504038.1 | 3 - 4 | 1115 | 70 | |
NM_182367.6 | NP_872167.2 | 3 - 4 | 971 | 70 | ||
GenBank mRNA | AY313178.1 | - | 3 - 4 | 1113 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |