Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dbl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072308.8 | NP_504709.1 | 3 - 4 | 609 | 107 | |
GenBank mRNA | AF004395.1 | - | 3 - 4 | 567 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |