Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dop-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028876.3 | NP_001024047.1 | 1 - 2 | 188 | 96 | |
NM_001269197.4 | NP_001256126.1 | 1 - 2 | 194 | 96 | ||
NM_001269198.4 | NP_001256127.1 | 1 - 2 | 189 | 96 | ||
NM_001373076.2 | NP_001360549.1 | 1 | 170 | 96 | ||
NM_001373077.2 | NP_001360548.1 | 1 | 170 | 96 | ||
NM_001373078.2 | NP_001360550.1 | 1 - 2 | 186 | 96 | ||
GenBank mRNA | AB125702.1 | - | 1 - 2 | 210 | 96 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |