Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lsy-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269216.3 | NP_001256145.1 | 5 - 6 | 1210 | 98 | |
NM_001269217.3 | NP_001256146.1 | 3 - 4 | 348 | 98 | ||
NM_001269218.3 | NP_001256147.1 | 6 - 7 | 1335 | 98 | ||
NM_001276918.3 | NP_001263847.1 | 5 - 6 | 1200 | 98 | ||
NM_001276919.3 | NP_001263848.1 | 3 - 4 | 348 | 98 | ||
NM_001276920.4 | NP_001263849.1 | 6 - 7 | 1335 | 98 | ||
NM_001373080.3 | NP_001359747.1 | 4 | 569 | 98 | ||
NM_001373081.2 | NP_001360180.1 | 4 | 550 | 98 | ||
NM_001373082.2 | NP_001359576.1 | 4 | 569 | 98 | ||
NM_001380733.2 | NP_001368168.1 | 4 - 5 | 498 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |