Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sulp-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073589.5 | NP_505990.3 | 10 - 11 | 1728 | 126 | |
GenBank mRNA | AY887911.1 | - | 10 - 11 | 1750 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |