Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shw-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001368471.3 | NP_001355445.1 | 4 - 5 | 317 | 72 | |
NM_001380790.1 | NP_001368707.1 | 4 - 5 | 308 | 72 | ||
NM_001380791.1 | NP_001366842.1 | 4 - 5 | 342 | 72 | ||
GenBank mRNA | DQ185514.1 | - | 3 - 4 | 254 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |