Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dct-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028706.7 | NP_001023877.1 | 4 - 5 | 395 | 130 | |
NM_001028707.6 | NP_001023878.1 | 4 - 5 | 401 | 130 | ||
NM_001028708.6 | NP_001023879.1 | 4 - 5 | 403 | 130 | ||
NM_001028709.5 | NP_001023880.1 | 4 - 5 | 403 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |