Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sec-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076129.6 | NP_508530.1 | 5 - 6 | 923 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |