Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acs-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373312.4 | NP_001360705.1 | 6 | 815 | 66 | |
NM_076592.9 | NP_508993.2 | 12 - 13 | 1927 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |