Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ilys-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029423.7 | NP_001024594.1 | 4 - 5 | 380 | 73 | |
NM_001313410.4 | NP_001300339.1 | 3 | 242 | 73 | ||
NR_132543.1 | 3 - 4 | 1059 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Caenorhabditis elegans Probe spans exons |