Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hum-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270229.4 | NP_001257158.1 | 6 - 7 | 1126 | 82 | |
NM_001270230.4 | NP_001257159.1 | 5 - 6 | 859 | 82 | ||
NM_001307862.4 | NP_001294791.1 | 6 - 7 | 1126 | 82 | ||
NM_001307863.4 | NP_001294792.1 | 5 - 6 | 859 | 82 | ||
NM_001373506.3 | NP_001360186.1 | 22 | 4732 | 82 | ||
NM_001373507.3 | NP_001359540.1 | 23 | 4796 | 82 | ||
NM_001383647.1 | 22 - 23 | 4809 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |