Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gei-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083303.5 | NP_001076772.1 | 23 - 24 | 3245 | 61 | |
NM_001083304.6 | NP_001076773.1 | 6 - 7 | 950 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |