Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270274.4 | NP_001257203.1 | 5 - 6 | 827 | 69 | |
NM_001270275.4 | NP_001257204.1 | 4 - 5 | 740 | 69 | ||
NM_001270276.4 | NP_001257205.1 | 1 - 2 | 299 | 69 | ||
NM_001381127.1 | NP_001366740.1 | 7 - 8 | 1447 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |