Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hlb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129713.4 | NP_001123185.1 | 4 - 5 | 685 | 71 | |
NM_001322647.4 | NP_001309707.1 | 8 | 857 | 71 | ||
NM_001322648.4 | NP_001309708.1 | 7 - 8 | 811 | 71 | ||
NM_001322649.4 | NP_001309709.1 | 6 - 7 | 739 | 71 | ||
NM_001322650.3 | NP_001309710.1 | 6 - 7 | 733 | 71 | ||
NM_001322653.5 | NP_001309713.1 | 2 | 274 | 71 | ||
NM_001322654.3 | NP_001309714.1 | 3 - 4 | 274 | 71 | ||
NM_001322657.3 | NP_001309717.1 | 3 - 4 | 238 | 71 | ||
NM_001322658.3 | NP_001309718.1 | 3 - 4 | 238 | 71 | ||
NM_001377622.2 | NP_001364846.1 | 5 - 6 | 535 | 71 | ||
NM_001383669.1 | 6 - 7 | 745 | 71 | |||
NM_001383671.1 | 6 - 7 | 760 | 71 | |||
NM_077910.5 | NP_510311.2 | 4 - 5 | 693 | 71 |
Set Membership: |
![]() ![]() ![]() ![]() |