Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ram-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078021.4 | NP_510422.2 | 13 - 14 | 1485 | 118 | |
GenBank mRNA | AF218866.1 | - | 13 - 14 | 1507 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |