Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: msp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062457.10 | NP_494858.1 | 1 | 240 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Both primers and probe map within a single exon |