Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mbd-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025841.4 | NP_001021012.1 | 3 - 4 | 534 | 70 | |
GenBank mRNA | DQ010405.1 | - | 3 - 4 | 556 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |