Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDNRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031632.1 | NP_001026802.1 | 4 - 5 | 897 | 106 | |
XM_005628830.3 | XP_005628887.1 | 5 - 6 | 1094 | 106 | ||
XM_014119229.2 | XP_013974704.1 | 5 - 6 | 1093 | 106 | ||
GenBank mRNA | AB183284.2 | - | 4 - 5 | 897 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Canis familiaris Probe spans exons |