Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003151.2 | NP_001003151.2 | 1 - 2 | 65 | 87 | |
GenBank mRNA | AF047047.1 | - | 1 - 2 | 50 | 87 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Canis familiaris Inventoried Probe spans exons |