Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1RN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003096.2 | NP_001003096.1 | 1 - 2 | 164 | 89 | |
XM_022404316.1 | XP_022260024.1 | 2 - 3 | 189 | 89 | ||
XM_022404317.1 | XP_022260025.1 | 2 - 3 | 402 | 89 | ||
GenBank mRNA | AF216526.1 | - | 1 - 2 | 130 | 89 | |
AY026462.2 | - | 1 - 2 | 158 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Inventoried Probe spans exons |