Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1CC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003033.1 | NP_001003033.1 | 742 | 102 | ||
XM_005636001.3 | XP_005636058.1 | 1020 | 102 | |||
GenBank mRNA | AF525130.1 | - | 742 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris |