Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003112.1 | NP_001003112.1 | 14 - 15 | 4202 | 71 | |
XM_005638170.3 | XP_005638227.1 | 16 - 17 | 4327 | 71 | ||
XM_005638171.3 | XP_005638228.1 | 16 - 17 | 4037 | 71 | ||
GenBank mRNA | X99145.2 | - | 4202 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |