Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: UACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003112.1 | NP_001003112.1 | 9 - 10 | 1159 | 116 | |
XM_005638170.3 | XP_005638227.1 | 9 - 10 | 1194 | 116 | ||
XM_005638171.3 | XP_005638228.1 | 9 - 10 | 904 | 116 | ||
GenBank mRNA | X99145.2 | - | 1159 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |