Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003431514.3 | XP_003431562.2 | 5 - 6 | 658 | 69 | |
XM_003431515.4 | XP_003431563.2 | 5 - 6 | 746 | 69 | ||
XM_005625978.3 | XP_005626035.1 | 5 - 6 | 745 | 69 | ||
XM_005625979.3 | XP_005626036.1 | 5 - 6 | 749 | 69 | ||
XM_005625980.3 | XP_005626037.1 | 5 - 6 | 671 | 69 | ||
XM_005625981.3 | XP_005626038.1 | 5 - 6 | 636 | 69 | ||
XM_005625982.3 | XP_005626039.1 | 5 - 6 | 604 | 69 | ||
XM_022424540.1 | XP_022280248.1 | 5 - 6 | 591 | 69 | ||
XM_857845.5 | XP_862938.3 | 5 - 6 | 741 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris Probe spans exons |