Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LCNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014116916.2 | XP_013972391.1 | 2 - 3 | 1253 | 69 | |
XM_014116919.2 | XP_013972394.1 | 2 - 3 | 1253 | 69 | ||
XM_022423534.1 | XP_022279242.1 | 2 - 3 | 1253 | 69 | ||
XM_022423535.1 | XP_022279243.1 | 2 - 3 | 1253 | 69 | ||
XM_022423537.1 | XP_022279245.1 | 2 - 3 | 1253 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |